Tisk…
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin))
Dodavatel: Biorbyt
BIRBORB43866-100EA 0 CZK
BIRBORB43866-100
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin))
Protilátky
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat