Tisk…

Anti-FAS Mouse Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB43958-100EA 0 CZK
BIRBORB43958-100
Anti-FAS Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR