Tisk…
Anti-SELL Mouse Monoclonal Antibody (FITC (Fluorescein))
Dodavatel: Biorbyt
BIRBORB43966-100EA 0 CZK
BIRBORB43966-100
Anti-SELL Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: SELL (selectin L)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human