Tisk…

Anti-PECAM1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB43990-100EA 0 CZK
BIRBORB43990-100
Anti-PECAM1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR