Tisk…
Anti-ITGA2B Mouse Monoclonal Antibody (Biotin)
Dodavatel: Biorbyt
BIRBORB43997-0.1EA 0 CZK
BIRBORB43997-0.1
Anti-ITGA2B Mouse Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human