Tisk…
Anti-HLA-DR Mouse Monoclonal Antibody (Biotin)
Dodavatel: Biorbyt
BIRBORB44065-0.1EA 0 CZK
BIRBORB44065-0.1
Anti-HLA-DR Mouse Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human