Tisk…

Anti-FCGR3A Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB44123-0.1EA 0 CZK
BIRBORB44123-0.1
Anti-FCGR3A Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR