Tisk…

Anti-NCAM1 Mouse Monoclonal Antibody (Peridinin Chlorophyll)

Dodavatel: Biorbyt
BIRBORB44170-100EA 0 CZK
BIRBORB44170-100
Anti-NCAM1 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Protilátky
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR