Tisk…

Anti-CD46 Mouse Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB44231-0.1EA 10970 CZK
BIRBORB44231-0.1
Anti-CD46 Mouse Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: CD46 (CD46 molecule, complement regulatory protein)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR