Tisk…
Anti-CD63 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Dodavatel: Biorbyt
BIRBORB44244-100EA 0 CZK
BIRBORB44244-100
Anti-CD63 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Protilátky
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human