Tisk…
Anti-TNFRSF8 Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB44260-0.1EA 0 CZK
BIRBORB44260-0.1
Anti-TNFRSF8 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF8 (tumor necrosis factor receptor superfamily, member 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human