Tisk…

Anti-CD8A Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44283-100EA 0 CZK
BIRBORB44283-100
Anti-CD8A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR