Tisk…

Anti-CD36 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44519-100EA 0 CZK
BIRBORB44519-100
Anti-CD36 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR