Tisk…

Anti-TUBB3 Mouse Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB44545-0.1EA 0 CZK
BIRBORB44545-0.1
Anti-TUBB3 Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: TUBB3 (tubulin, beta 3 class III)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR