Tisk…

Anti-ANPEP Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44586-100EA 0 CZK
BIRBORB44586-100
Anti-ANPEP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR