Tisk…

Anti-CD3 Hamster Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44593-0.1EA 0 CZK
BIRBORB44593-0.1
Anti-CD3 Hamster Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR