Tisk…

Anti-FCGR1A Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB44627-100EA 0 CZK
BIRBORB44627-100
Anti-FCGR1A Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR