Tisk…
Anti-PDGFRA Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB44633-0.1EA 0 CZK
BIRBORB44633-0.1
Anti-PDGFRA Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PDGFRA (platelet-derived growth factor receptor, alpha polypeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human