Tisk…

Anti-PDGFRA Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44636-100EA 0 CZK
BIRBORB44636-100
Anti-PDGFRA Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PDGFRA (platelet-derived growth factor receptor, alpha polypeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR