Tisk…

Anti-CD163 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Dodavatel: Biorbyt
BIRBORB44714-100EA 0 CZK
BIRBORB44714-100
Anti-CD163 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Protilátky
Type: Primary
Antigen: CD163 (CD163 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR