Tisk…

Anti-TCR alpha Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44730-0.1EA 0 CZK
BIRBORB44730-0.1
Anti-TCR alpha Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TCR alpha
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR