Tisk…

Anti-CSK Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB44766-0.1EA 0 CZK
BIRBORB44766-0.1
Anti-CSK Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CSK (c-src tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR