Tisk…

Anti-ZAP70 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB44783-0.1EA 0 CZK
BIRBORB44783-0.1
Anti-ZAP70 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ZAP70 (zeta-chain (TCR) associated protein kinase 70kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR