Tisk…

Anti-LAT2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB44787-0.1EA 0 CZK
BIRBORB44787-0.1
Anti-LAT2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LAT2 (linker for activation of T cells family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR