Tisk…

Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB44797-0.1EA 0 CZK
BIRBORB44797-0.1
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR