Tisk…

Anti-ITGB2 Rat Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44829-0.1EA 0 CZK
BIRBORB44829-0.1
Anti-ITGB2 Rat Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR