Tisk…

Anti-IL2RA Rat Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB44836-0.1EA 0 CZK
BIRBORB44836-0.1
Anti-IL2RA Rat Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR