Tisk…

Anti-IgG Goat Polyclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB47919-500EA 0 CZK
BIRBORB47919-500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR