Tisk…

Anti-GCLC Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB47959-100EA 0 CZK
BIRBORB47959-100
Anti-GCLC Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: GCLC (glutamate-cysteine ligase, catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR