Tisk…

Anti-CASP10 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB47980-100EA 0 CZK
BIRBORB47980-100
Anti-CASP10 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: CASP10 (caspase 10, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR