Tisk…

Anti-CCL11 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB47998-100EA 0 CZK
BIRBORB47998-100
Anti-CCL11 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: CCL11 (chemokine (C-C motif) ligand 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR