Tisk…

Anti-ADAM10 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48002-100EA 0 CZK
BIRBORB48002-100
Anti-ADAM10 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ADAM10 (ADAM metallopeptidase domain 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR