Tisk…

Anti-BCL3 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB48006-100EA 0 CZK
BIRBORB48006-100
Anti-BCL3 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: BCL3 (B-cell CLL/lymphoma 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR