Tisk…

Anti-CDC25A Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48012-100EA 0 CZK
BIRBORB48012-100
Anti-CDC25A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR