Tisk…

Anti-CLDN7 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB48333-100EA 0 CZK
BIRBORB48333-100
Anti-CLDN7 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: CLDN7 (claudin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR