Tisk…
Anti-LGALS3 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB48437-100EA 0 CZK
BIRBORB48437-100
Anti-LGALS3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LGALS3 (lectin, galactoside-binding, soluble, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat