Tisk…

Anti-VTN Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48616-200EA 0 CZK
BIRBORB48616-200
Anti-VTN Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: VTN (vitronectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR