Tisk…

Anti-GDF5 Chicken polyclonal antibody

Dodavatel: Biorbyt
BIRBORB48621-200EA 0 CZK
BIRBORB48621-200
Anti-GDF5 Chicken polyclonal antibody
Protilátky
Type: Primary
Antigen: GDF5 (growth differentiation factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR