Tisk…

Anti-AGRP Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48629-200EA 0 CZK
BIRBORB48629-200
Anti-AGRP Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:64000; Western Blot: 1:1000

Type: Primary
Antigen: AGRP (agouti related protein homolog (mouse))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR