Tisk…

Anti-Doublesex Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB48636-200EA 0 CZK
BIRBORB48636-200
Anti-Doublesex Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Doublesex
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Drosophila
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR