Tisk…

Anti-TG Chicken Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48637-200EA 0 CZK
BIRBORB48637-200
Anti-TG Chicken Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TG (thyroglobulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR