Tisk…

Anti-PCT Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48641-200EA 0 CZK
BIRBORB48641-200
Anti-PCT Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:2000-4000; Western Blot: 1:1000-2000

Type: Primary
Antigen: PCT
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR