Tisk…

Anti-GCG Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB48649-200EA 0 CZK
BIRBORB48649-200
Anti-GCG Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: GCG (glucagon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR