Tisk…

Anti-Tau 441 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB48681-200EA 0 CZK
BIRBORB48681-200
Anti-Tau 441 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Tau 441
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR