Tisk…

Anti-ERK Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48685-100EA 0 CZK
BIRBORB48685-100
Anti-ERK Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ERK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR