Tisk…

Anti-Inhibin A beta Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB48760-200EA 0 CZK
BIRBORB48760-200
Anti-Inhibin A beta Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Inhibin A beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR