Tisk…
Anti-Cyfra21-1 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB48781-200EA 0 CZK
BIRBORB48781-200
Anti-Cyfra21-1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Cyfra21-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human