Tisk…

Anti-Endorphin B Rabbit monoclonal antibody

Dodavatel: Biorbyt
BIRBORB48876-200EA 0 CZK
BIRBORB48876-200
Anti-Endorphin B Rabbit monoclonal antibody
Protilátky
Type: Primary
Antigen: Endorphin B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR