Tisk…

Anti-CKB Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB48905-200EA 0 CZK
BIRBORB48905-200
Anti-CKB Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: CKB (creatine kinase, brain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR