Tisk…

Anti-TNFRSF1B Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB49012-200EA 0 CZK
BIRBORB49012-200 BIRBORB49012-100
Anti-TNFRSF1B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF1B (tumor necrosis factor receptor superfamily, member 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR